Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-methylbenzamide ≥95% research grade

Supplier: AOB CHEM USA
Warning

26345-1G 26345-25G 26345-5G
76912-340EA 76.39 USD InStock
76912-340 76912-342 76912-344
3-Bromo-5-methylbenzamide ≥95% research grade
3-Bromo-5-methylbenzamide


Formula: C₈H₈BrNO
MW: 214.059 g/mol
Storage Temperature: Ambient
CAS Number: 1007578-82-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty