Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-(trifluoromethyl)benzamide ≥97% research grade

Supplier: AOB CHEM USA
Warning

26357-1G 26357-25G 26357-500MG 26357-5G
76912-352EA 126.25 USD InStock
76912-352 76912-354 76912-356 76912-358
4-Bromo-2-(trifluoromethyl)benzamide ≥97% research grade
4-Bromo-2-(trifluoromethyl)benzamide


Formula: C₈H₅BrF₃NO
MW: 268.031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27939894
CAS Number: 123524-61-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty