Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Formyl-5-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

26914-1G 26914-25G 26914-500MG 26914-5G
76912-564EA 277.96 USD InStock
76912-564 76912-566 76912-568 76912-570
3-Formyl-5-methylbenzoic acid ≥97%
3-Formyl-5-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD20645804
CAS Number: 4481-27-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty