Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorophenyl)benzonitrile ≥97% research grade

Supplier: AOB CHEM USA
Warning

29069-25G 29069-5G
76913-126EA 580.32 USD InStock
76913-126 76913-128
4-(4-Fluorophenyl)benzonitrile ≥97% research grade
4-(4-Fluorophenyl)benzonitrile


Formula: C₁₃H₈FN
Storage Temperature: Ambient
MDL Number: MFCD05981088
CAS Number: 10540-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty