Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dimethyl-2-methoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

29678-1G 29678-25G 29678-500MG 29678-5G
76913-360EA 381.93 USD InStock
76913-360 76913-362 76913-364 76913-366
4,5-Dimethyl-2-methoxyphenylboronic acid ≥97%
4,5-Dimethyl-2-methoxyphenylboronic acid


Formula: C₉H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28795053
CAS Number: 2121511-71-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty