Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[(tert-Butoxycarbonyl)amino]isonicotinic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

29756-1G 29756-25G 29756-5G
76913-400EA 81.69 USD InStock
76913-400 76913-402 76913-404
3-[(tert-Butoxycarbonyl)amino]isonicotinic acid ≥95% research grade
3-[(tert-Butoxycarbonyl)amino]isonicotinic acid


Formula: C₁₁H₁₄N₂O₄
MW: 238.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03427723
CAS Number: 179024-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty