Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-4'-chlorobiphenyl ≥97%

Supplier: AOB CHEM USA
Warning

30156-1G 30156-25G 30156-500MG 30156-5G
76913-486EA 224.92 USD InStock
76913-486 76913-488 76913-490 76913-492
4-Acetyl-4'-chlorobiphenyl ≥97%
4-Acetyl-4'-chlorobiphenyl


Formula: C₁₄H₁₁ClO
Storage Temperature: Refrigerator
MDL Number: MFCD04039062
CAS Number: 5002-07-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty