Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluorophenyl methyl sulfone ≥97%

Supplier: AOB CHEM USA
Warning

30899-1G 30899-25G 30899-5G
76913-654EA 79.57 USD InStock
76913-654 76913-656 76913-658
5-Bromo-2-fluorophenyl methyl sulfone ≥97%
5-Bromo-2-fluorophenyl methyl sulfone
Precaución: for Research Use Only


Formula: C₇H₆BrFO₂S
MW: 253.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458078
CAS Number: 914636-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty