Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-2-fluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

31054-1G 31054-25G 31054-500MG 31054-5G
76913-746EA 201.58 USD InStock
76913-746 76913-748 76913-750 76913-752
3,5-Dichloro-2-fluorobenzoic acid ≥97%
3,5-Dichloro-2-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Cl₂FO₂
MW: 209.002 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04091131
CAS Number: 665022-07-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty