Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,5-dimethylbenzyl alcohol ≥97% research grade

Supplier: AOB CHEM USA
Warning

31158-1G 31158-25G 31158-500MG 31158-5G
76913-824EA 201.58 USD InStock
76913-824 76913-826 76913-828 76913-830
4-Bromo-2,5-dimethylbenzyl alcohol ≥97% research grade
4-Bromo-2,5-dimethylbenzyl alcohol


Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD18905464
CAS Number: 952303-55-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty