Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,3-dichlorobenzoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

31319-1G 31319-25G 31319-500MG 31319-5G
76913-908EA 154.9 USD InStock
76913-908 76913-910 76913-912 76913-914
4-Bromo-2,3-dichlorobenzoic acid ≥95% research grade
4-Bromo-2,3-dichlorobenzoic acid


Formula: C₇H₃BrCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD19686181
CAS Number: 1257517-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty