Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dimethylamino)-5-fluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

31405-1G 31405-25G 31405-500MG 31405-5G
76913-938EA 215.37 USD InStock
76913-938 76913-940 76913-942 76913-944
3-(Dimethylamino)-5-fluorobenzoic acid ≥97%
3-(Dimethylamino)-5-fluorobenzoic acid


Formula: C₉H₁₀FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD26791453
CAS Number: 1350539-93-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty