Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-6-chloropyridine-3-boronic acid, pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

31642-1G 31642-25G 31642-500MG 31642-5G
76914-004EA 183.54 USD InStock
76914-004 76914-006 76914-008 76914-010
5-Amino-6-chloropyridine-3-boronic acid, pinacol ester ≥97%
5-Amino-6-chloropyridine-3-boronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₆BClN₂O₂
MW: 254.521 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09260487
CAS Number: 1073354-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty