Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hydroxymethyl)-2-methylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

32076-1G 32076-500MG 32076-5G
76914-088EA 524.09 USD InStock
76914-088 76914-090 76914-092
3-(Hydroxymethyl)-2-methylphenylboronic acid ≥97%
3-(Hydroxymethyl)-2-methylphenylboronic acid


Formula: C₈H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28969493
CAS Number: 2121514-26-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty