Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluoro-4-methoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

32259-1G 32259-25G 32259-500MG 32259-5G
76914-148EA 290.69 USD InStock
76914-148 76914-150 76914-152 76914-154
3-Chloro-2-fluoro-4-methoxybenzoic acid ≥97% research grade
3-Chloro-2-fluoro-4-methoxybenzoic acid


Formula: C₈H₆ClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28640403
CAS Number: 1554885-43-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty