Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,5-dimethoxy-benzoic acid methyl ester ≥95%

Supplier: AOB CHEM USA
Warning

32316-1G 32316-25G 32316-500MG 32316-5G
76914-192EA 290.69 USD InStock
76914-192 76914-194 76914-196 76914-198
4-Bromo-2,5-dimethoxy-benzoic acid methyl ester ≥95%
4-Bromo-2,5-dimethoxy-benzoic acid methyl ester


Formula: C₁₀H₁₁BrO₄
Storage Temperature: Refrigerator
MDL Number: MFCD28738428
CAS Number: 855749-57-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty