Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,4-dimethylphenol ≥97%

Supplier: AOB CHEM USA
Warning

32632-1G 32632-25G 32632-500MG 32632-5G
76914-368EA 439.22 USD InStock
76914-368 76914-370 76914-372 76914-374
3-Chloro-2,4-dimethylphenol ≥97%
3-Chloro-2,4-dimethylphenol


Formula: C₈H₉ClO
Storage Temperature: Refrigerator
MDL Number: MFCD22414681
CAS Number: 56680-84-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty