Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Chloro-4-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOB CHEM USA
Warning

32639-1G 32639-25G 32639-500MG 32639-5G
76914-384EA 183.54 USD InStock
76914-384 76914-386 76914-388 76914-390
2-(3-Chloro-4-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(3-Chloro-4-ethoxy-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane


Formula: C₁₄H₁₉BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD24368960
CAS Number: 1668474-08-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty