Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-trifluoromethylpyridine-5-boronic acid ≥95%

Supplier: AOB CHEM USA
Warning

32740-1G 32740-500MG 32740-5G
76914-448EA 528.33 USD InStock
76914-448 76914-450 76914-452
3-Methyl-2-trifluoromethylpyridine-5-boronic acid ≥95%
3-Methyl-2-trifluoromethylpyridine-5-boronic acid


Formula: C₇H₇BF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22491393
CAS Number: 1889221-03-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty