Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluoro-4-(methoxymethoxy)benzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

32804-1G 32804-25G 32804-500MG 32804-5G
76914-474EA 225.98 USD InStock
76914-474 76914-476 76914-478 76914-480
3-Chloro-5-fluoro-4-(methoxymethoxy)benzoic acid ≥97% research grade
3-Chloro-5-fluoro-4-(methoxymethoxy)benzoic acid


Formula: C₉H₈ClFO₄
MW: 234.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619444
CAS Number: 2179038-39-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty