Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-chloro-2-methoxybenzoate ≥97% research grade

Supplier: AOB CHEM USA
Warning

32945-1G 32945-25G 32945-5G
76914-502EA 62.6 USD InStock
76914-502 76914-504 76914-506
Methyl 3-chloro-2-methoxybenzoate ≥97% research grade
Methyl 3-chloro-2-methoxybenzoate


Formula: C₉H₉ClO₃
MW: 200.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547158
CAS Number: 92992-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty