Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxy-4-fluorophenol ≥97% research grade

Supplier: AOB CHEM USA
Warning

33099-1G 33099-25G 33099-500MG 33099-5G
76914-566EA 153.84 USD InStock
76914-566 76914-568 76914-570 76914-572
3-Benzyloxy-4-fluorophenol ≥97% research grade
3-Benzyloxy-4-fluorophenol


Formula: C₁₃H₁₁FO₂
MW: 218.224 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30723167
CAS Number: 1256550-88-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty