Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluoro-5-trifluoromethylphenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

33500-1G 33500-25G 33500-500MG 33500-5G
76914-704EA 128.37 USD InStock
76914-704 76914-706 76914-708 76914-710
3-Bromo-2-fluoro-5-trifluoromethylphenylboronic acid pinacol ester ≥97%
3-Bromo-2-fluoro-5-trifluoromethylphenylboronic acid pinacol ester


Formula: C₁₃H₁₄BBrF₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD31806029
CAS Number: 1799485-19-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty