Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluoro-2-hydroxyphenylboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

33542-1G 33542-25G 33542-500MG 33542-5G
76914-766EA 171.87 USD InStock
76914-766 76914-768 76914-770 76914-772
3-Chloro-5-fluoro-2-hydroxyphenylboronic acid ≥95%
3-Chloro-5-fluoro-2-hydroxyphenylboronic acid


Formula: C₆H₅BClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22414782
CAS Number: 2121515-30-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty