Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,5-dichlorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

33654-1G 33654-25G 33654-500MG 33654-5G
76914-820EA 159.14 USD InStock
76914-820 76914-822 76914-824 76914-826
3-Bromo-2,5-dichlorobenzoic acid ≥97%
3-Bromo-2,5-dichlorobenzoic acid


Formula: C₇H₃BrCl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07780678
CAS Number: 855202-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty