Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-5-methoxyphenylboronic acid pinacol eater ≥97% research grade

Supplier: AOB CHEM USA
Warning

33804-1G 33804-25G 33804-500MG 33804-5G
76914-888EA 320.4 USD InStock
76914-888 76914-890 76914-892 76914-894
3-Ethoxy-5-methoxyphenylboronic acid pinacol eater ≥97% research grade
3-Ethoxy-5-methoxyphenylboronic acid pinacol eater


Formula: C₁₅H₂₃BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD19237211
CAS Number: 2096995-78-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty