Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-phenylpyridine ≥95%

Supplier: AOB CHEM USA
Warning

34353-1G 34353-25G 34353-500MG 34353-5G
76915-068EA 126.25 USD InStock
76915-068 76915-070 76915-072 76915-074
3-Bromo-2-phenylpyridine ≥95%
3-Bromo-2-phenylpyridine


Formula: C₁₁H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD04114116
CAS Number: 91182-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty