Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzoxy)-5-chlorophenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

34460-1G 34460-25G 34460-5G
76915-084EA 99.73 USD InStock
76915-084 76915-086 76915-088
3-(Benzoxy)-5-chlorophenylboronic acid ≥97% research grade
3-(Benzoxy)-5-chlorophenylboronic acid


Formula: C₁₃H₁₂BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16660283
CAS Number: 1256345-82-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty