Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-phenylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

34793-1G 34793-25G 34793-500MG 34793-5G
76915-222EA 409.51 USD InStock
76915-222 76915-224 76915-226 76915-228
3-Chloro-4-phenylbenzaldehyde ≥97% research grade
3-Chloro-4-phenylbenzaldehyde


Formula: C₁₃H₉ClO
MW: 216.6630 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30476460
CAS Number: 57592-44-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty