Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dibromo-2-fluorobenzoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

36815-1G 36815-25G 36815-500MG 36815-5G
76915-670EA 173.99 USD InStock
76915-670 76915-672 76915-674 76915-676
3,4-Dibromo-2-fluorobenzoic acid ≥95% research grade
3,4-Dibromo-2-fluorobenzoic acid


Formula: C₇H₃Br₂FO₂
MW: 297.904 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28784653
CAS Number: 1804416-86-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty