Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)-6-bromo-2-chlorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

37220-1G 37220-25G 37220-500MG 37220-5G
76915-744EA 209 USD InStock
76915-744 76915-746 76915-748 76915-750
3-(Benzyloxy)-6-bromo-2-chlorobenzoic acid ≥97% research grade
3-(Benzyloxy)-6-bromo-2-chlorobenzoic acid


Formula: C₁₄H₁₀BrClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632166
CAS Number: 2379322-11-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty