Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-4-fluoro-5-methylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

37294-1G 37294-25G 37294-500MG 37294-5G
76915-758EA 421.18 USD InStock
76915-758 76915-760 76915-762 76915-764
3-Ethoxy-4-fluoro-5-methylbenzoic acid ≥97% research grade
3-Ethoxy-4-fluoro-5-methylbenzoic acid


Formula: C₁₀H₁₁FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632189
CAS Number: 2383799-38-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty