Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-6-fluorobenzonitrile ≥97% research grade

Supplier: AOB CHEM USA
Warning

37701-1G 37701-25G 37701-500MG 37701-5G
76915-800EA 266.29 USD InStock
76915-800 76915-802 76915-804 76915-806
3-Bromo-2-chloro-6-fluorobenzonitrile ≥97% research grade
3-Bromo-2-chloro-6-fluorobenzonitrile


Formula: C₇H₂BrClFN
MW: 234.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28348546
CAS Number: 1421620-33-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty