Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-3,5-difluorobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

37985-1G 37985-25G 37985-500MG 37985-5G
76915-858EA 242.95 USD InStock
76915-858 76915-860 76915-862 76915-864
4-(Benzyloxy)-3,5-difluorobenzoic acid ≥97% research grade
4-(Benzyloxy)-3,5-difluorobenzoic acid


Formula: C₁₄H₁₀F₂O₃
MW: 264.224 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18545674
CAS Number: 1408143-67-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty