Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzoic acid hydrazide ≥95%

Supplier: AOB CHEM USA
Warning

38536-25G 38536-5G
76915-974EA 212.18 USD InStock
76915-974 76915-976
4-(Trifluoromethoxy)benzoic acid hydrazide ≥95%
4-(Trifluoromethoxy)benzoic acid hydrazide
Precaución: for Research Use Only


Formula: C₈H₇F₃N₂O₂
MW: 220.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204234
CAS Number: 175277-18-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty