Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(methylsulfonyl)benzeneboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

39159-1G 39159-25G 39159-500MG 39159-5G
76916-016EA 195.21 USD InStock
76916-016 76916-018 76916-020 76916-022
3-Fluoro-4-(methylsulfonyl)benzeneboronic acid ≥97%
3-Fluoro-4-(methylsulfonyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BFO₄S
Storage Temperature: Ambient
MDL Number: MFCD09878323
CAS Number: 648904-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty