Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-5-ethoxy-4-fluorophenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

40353-1G 40353-25G 40353-500MG 40353-5G
76916-154EA 202.64 USD InStock
76916-154 76916-156 76916-158 76916-160
(3-Chloro-5-ethoxy-4-fluorophenyl)boronic acid ≥97% research grade
(3-Chloro-5-ethoxy-4-fluorophenyl)boronic acid


Formula: C₈H₉BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD20441834
CAS Number: 2096339-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty