Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-nitro-4-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97% research grade

Supplier: AOB CHEM USA
Warning

41104-1G 41104-25G 41104-500MG 41104-5G
76916-222EA 138.98 USD InStock
76916-222 76916-224 76916-226 76916-228
3-nitro-4-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97% research grade
3-nitro-4-(trifluoromethoxy)phenylboronic acid, pinacol ester


Formula: C₁₃H₁₅BF₃NO₅
MW: 333.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21333035
CAS Number: 2096336-45-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty