Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-2,4-dimethylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

49749-1G 49749-500MG 49749-5G
76916-716EA 795.68 USD InStock
76916-716 76916-718 76916-720
3-Methoxy-2,4-dimethylbenzoic acid ≥97% research grade
3-Methoxy-2,4-dimethylbenzoic acid


Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28794947
CAS Number: 246877-25-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty