Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)-2-methoxybenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

52490-1G 52490-25G 52490-500MG 52490-5G
76916-878EA 215.37 USD InStock
76916-878 76916-880 76916-882 76916-884
4-(Benzyloxy)-2-methoxybenzoic acid ≥97% research grade
4-(Benzyloxy)-2-methoxybenzoic acid


Formula: C₁₅H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD06203640
CAS Number: 85607-79-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty