Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-tert-butylbenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

52983-1G 52983-25G 52983-5G
76916-936EA 88.06 USD InStock
76916-936 76916-938 76916-940
3-Bromo-5-tert-butylbenzoic acid ≥97% research grade
3-Bromo-5-tert-butylbenzoic acid


Formula: C₁₁H₁₃BrO₂
MW: 257.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07357307
CAS Number: 794465-45-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty