Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzylthio)benzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

53062-1G 53062-25G 53062-5G
76916-966EA 84.88 USD InStock
76916-966 76916-968 76916-970
4-(Benzylthio)benzoic acid ≥95%
4-(Benzylthio)benzoic acid


Formula: C₁₄H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD06804476
CAS Number: 22855-95-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty