Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Piperidin-1-ylbenzoic acid ≥95%

Supplier: AOB CHEM USA
Warning

53723-1G 53723-25G 53723-500MG 53723-5G
76917-068EA 144.29 USD InStock
76917-068 76917-070 76917-072 76917-074
3-Piperidin-1-ylbenzoic acid ≥95%
3-Piperidin-1-ylbenzoic acid


Formula: C₁₂H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD06659081
CAS Number: 77940-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty