Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromobenzo[b]thiophene-2-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

54344-1G 54344-25G 54344-500MG 54344-5G
76917-124EA 135.8 USD InStock
76917-124 76917-126 76917-128 76917-130
3-Bromobenzo[b]thiophene-2-carboxylic acid ≥97%
3-Bromobenzo[b]thiophene-2-carboxylic acid


Formula: C₉H₅BrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD466543
CAS Number: 29174-66-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty