Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluoro-6-iodobenzoic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

55948-1G 55948-25G 55948-500MG 55948-5G
76917-228EA 173.99 USD InStock
76917-228 76917-230 76917-232 76917-234
3-Bromo-2-fluoro-6-iodobenzoic acid ≥97% research grade
3-Bromo-2-fluoro-6-iodobenzoic acid


Formula: C₇H₃BrFIO₂
MW: 344.904 g/mol
Storage Temperature: Ambient
CAS Number: 217816-54-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty