Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4- Bromo-5-ethyl-1-methyl-1H-pyrazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

57070-1G 57070-25G 57070-500MG 57070-5G
76917-314EA 245.07 USD InStock
76917-314 76917-316 76917-318 76917-320
4- Bromo-5-ethyl-1-methyl-1H-pyrazole ≥97% research grade
4- Bromo-5-ethyl-1-methyl-1H-pyrazole


Formula: C₆H₉BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD22548406
CAS Number: 1393728-48-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty