Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-4-(methylthio)benzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

60120-1G 60120-25G 60120-500MG 60120-5G
76917-468EA 165.51 USD InStock
76917-468 76917-470 76917-472 76917-474
3-Methoxy-4-(methylthio)benzaldehyde ≥97% research grade
3-Methoxy-4-(methylthio)benzaldehyde


Formula: C₉H₁₀O₂S
Storage Temperature: Ambient
CAS Number: 68885-46-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty