Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(6-Chloro-2-pyridinyl)morpholine ≥97% research grade

Supplier: AOB CHEM USA
Warning

61164-1G 61164-25G 61164-500MG 61164-5G
76917-514EA 296 USD InStock
76917-514 76917-516 76917-518 76917-520
4-(6-Chloro-2-pyridinyl)morpholine ≥97% research grade
4-(6-Chloro-2-pyridinyl)morpholine


Formula: C₉H₁₁ClN₂O
MW: 198.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848425
CAS Number: 330682-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty