Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methoxymethyl)benzoic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

65775-1G 65775-25G 65775-500MG 65775-5G
76917-768EA 124.13 USD InStock
76917-768 76917-770 76917-772 76917-774
3-(Methoxymethyl)benzoic acid ≥95% research grade
3-(Methoxymethyl)benzoic acid


Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD01871355
CAS Number: 32194-76-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty