Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-methyl-5-nitrobenzamide ≥97% research grade

Supplier: AOB CHEM USA
Warning

73240-1G 73240-25G 73240-500MG 73240-5G
76917-922EA 416.94 USD InStock
76917-922 76917-924 76917-926 76917-928
3-Bromo-4-methyl-5-nitrobenzamide ≥97% research grade
3-Bromo-4-methyl-5-nitrobenzamide


Formula: C₈H₇BrN₂O₃
Storage Temperature: Ambient
CAS Number: 1000343-37-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty